Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2107938

Sigma-Aldrich

Anti-CCL5 antibody produced in rabbit

IgG fraction of antiserum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

10kDa

species reactivity

rat, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... CCL5(20304)

Immunogen

Synthetic peptide directed towards the middle region of mouse CCL5

Biochem/physiol Actions

Recently, it has been shown that genetic polymorphisms can result in diminished expression of CCL5, which results in increased susceptibility to and progression of infectious diseases. CCL5, together with Th cytokine mRNA expression, is temporally up-regulated during pneumococcal carriage. CCL5 is an essential factor for the induction and maintenance of protective pneumococcal immunity

Sequence

Synthetic peptide located within the following region: YGSDTTPCCFAYLSLELPRAHVKEYFYTSSKCSNLAVVFVTRRNRQVCAN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Fe Lanfranco et al.
Frontiers in neuroanatomy, 11, 137-137 (2018-01-30)
Chemokine (C-C motif) ligand 5 (CCL5) belongs to a group of chemokines that play a role in the peripheral immune system, mostly as chemoattractant molecules, and mediate tactile allodynia. In the central nervous system (CNS), CCL5 and its receptors have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service