Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2102661

Sigma-Aldrich

Anti-USP18 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ISG43, Anti-UBP43, Anti-Ubiquitin specific peptidase 18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

43 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP18(11274)

Immunogen

Synthetic peptide directed towards the N terminal region of human USP18

Biochem/physiol Actions

USP18, a member of the deubiquitinating protease family of enzymes, removes ubiquitin adducts from a broad range of protein substrates.USP18, a member of the deubiquitinating protease family of enzymes, removes ubiquitin adducts from a broad range of protein substrates.[supplied by OMIM]. Sequence Note: The RefSeq transcript and protein were derived from genomic sequence to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on alignments. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-232 AC008079.24 80964-81195 233-495 AC008079.24 88531-88793 496-592 AC008079.24 91145-91241 593-738 AC008079.24 92763-92908 739-818 AC008079.24 98228-98307 819-965 AC008079.24 98863-99009 966-1061 AC008079.24 100817-100912 1062-1229 AC008079.24 101726-101893 1230-1361 AC008079.24 104123-104254 1362-1411 AC008079.24 104766-104815 1412-2037 AC008079.24 107745-108370

Sequence

Synthetic peptide located within the following region: MQDSRQKAVRPLELAYCLQKCNVPLFVQHDAAQLYLKLWNLIKDQITDVH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service