Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2101692

Sigma-Aldrich

Anti-OSBPL8 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp686A11164, Anti-MGC126578, Anti-MGC133203, Anti-MST120, Anti-Oxysterol binding protein-like 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

97 kDa

species reactivity

bovine, human, pig, rabbit, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OSBPL8(114882)

General description

The previously assigned protein identifier Q68D75 has been merged into Q9BZF1. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the N terminal region of human OSBPL8

Biochem/physiol Actions

OSBPL8 is a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, OSBPL8 contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding different isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: SQRQGKEAYPTPTKDLHQPSLSPASPHSQGFERGKEDISQNKDESSLSMS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service