Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB2101578

Sigma-Aldrich

Anti-NFATC3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-NFAT4, Anti-NFATX, Anti-Nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

115 kDa

species reactivity

dog, mouse, human, horse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFATC3(4775)

General description

The nuclear factor of activated T cells 3 (NFATc3)/nuclear factor of activated T cells 4 (NFAT4) belongs to the NFAT family. It is expressed at high levels in the thymus. NFATc3 has a rel similarity domain (RSD) and the SP repeat region, characterized by the repeated motif SPxxSPxxSPrxsxx(D/E)(D/E)swl. The NFATc3 gene is located on human chromosome 16.

Immunogen

Synthetic peptide directed towards the N terminal region of human NFATC3

Application

Anti-NFATC3 antibody produced in rabbit has been used in western blot analysis (1:1000).

Biochem/physiol Actions

The nuclear factor of activated T cells 3 (NFATc3)/nuclear factor of activated T cells 4 (NFAT4) exhibits either pro-tumor or anti-tumor effects. It can block the proliferation and migration abilities of hepatoma cells. NFATc3 aids in reducing hepatitis B virus (HBV) replication. It can activate transcription. NFAT4 is necessary to drive the expression of the human polyomavirus JC virus (JCV) gene in glial cells.

Sequence

Synthetic peptide located within the following region: AVFPFQYCVETDIPLKTRKTSEDQAAILPGKLELCSDDQGSLSPARETSI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaobin Zao et al.
Oncoimmunology, 10(1), 1869388-1869388 (2021-02-02)
Nuclear factor of activated T cells 3 (NFATc3) has been reported to upregulate type I interferons (IFNs) expression, and the abnormal expression and activation of NFATc3 were closely related to tumorigenesis. However, the potential function of NFATc3 in hepatitis B
S N Ho et al.
The Journal of biological chemistry, 270(34), 19898-19907 (1995-08-25)
Signals transduced by the T cell antigen receptor (TCR) regulate developmental transitions in the thymus and also mediate the immunologic activation of mature, peripheral T cells. In both cases TCR stimulation leads to the assembly of the NFAT transcription complex
Kate Manley et al.
Journal of virology, 80(24), 12079-12085 (2006-10-13)
The human polyomavirus JC virus (JCV) infects 70% of the population worldwide. In immunosuppressed patients, JCV infection can lead to progressive multifocal leukoencephalopathy (PML), a fatal demyelinating disease of the central nervous system (CNS). The majority of PML cases occur
Patrick Nasarre et al.
Cancers, 13(11) (2021-06-03)
Secreted frizzled-related protein 2 (SFRP2) promotes the migration/invasion of metastatic osteosarcoma (OS) cells and tube formation by endothelial cells. However, its function on T-cells is unknown. We hypothesized that blocking SFRP2 with a humanized monoclonal antibody (hSFRP2 mAb) can restore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service