Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1413292

Sigma-Aldrich

Monoclonal Anti-CLDN6 antibody produced in mouse

clone 1P8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CLDN6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800

conjugate

unconjugated

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1P8, monoclonal

form

buffered aqueous solution

mol wt

23.3 kDa

species reactivity

human

concentration

~0.3-1 μg/μL (Please refer to lot specific concentration)

technique(s)

ELISA: suitable

isotype

IgG2b, kappa

Protein ID accession no.

NM_021195.3

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... CLDN6(9074)

General description

Mouse monoclonal antibody raised against a full-length recombinant CLDN6.

Immunogen

CLDN6 (NP_067018.1, 1 a.a. ~ 220 a.a) full-length recombinant protein with GST tag.

Sequence

MASAGMQILGVVLTLLGWVNGLVSCALPMWKVTAFIGNSIVVAQVVWEGLWMSCVVQSTGQMQCKVYDSLLALPQDLQAARALCVIALLVALFGLLVYLAGAKCTTCVEEKDSKARLVLTSGIVFVISGVLTLIPVCWTAHAVIRDFYNPLVAEAQKRELGASLYLGWAASGLLLLGGGLLCCTCPSGGSQGPSHYMARYSTSAPAISRGPSEYPTKNYV

Physical form

Solution in phosphate buffered saline, pH 7.4

Preparation Note

Purification by Protein A sepharose

Storage and Stability

Storage Instruction: Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service