Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB1413281

Sigma-Aldrich

Monoclonal Anti-RLF antibody produced in mouse

new

clone 2G2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

RLF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800

conjugate

unconjugated

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2G2, monoclonal

form

buffered aqueous solution

mol wt

37.73 kDa

species reactivity

human

concentration

~0.3-1 μg/μL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2a, kappa

Protein ID accession no.

NM_012421

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... RLF(6018)

General description

Mouse monoclonal antibody raised against a partial recombinant RLF.

Immunogen

RLF (NP_036553.1, 1805 a.a. ~ 1913 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence

SNDLTGNVVANNMVNDSEPEVDIPHSSSDSTIHENLTAIPPLIVAETTTVPSLENLRVVLDKALTDCGELALKQLHYLRPVVVLERSKFSTPILDLFPTKKTDELCVGS

Physical form

Solution in phosphate buffered saline, pH 7.4

Preparation Note

Purification by Protein A sepharose

Storage and Stability

Storage Instruction: Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L M Bourke et al.
Differentiation; research in biological diversity, 94, 8-20 (2016-12-09)
Recently we reported that Rearranged L-Myc Fusion, RLF, acts as an epigenetic modifier maintaining low levels of DNA methylation at CpG island shores and enhancers across the genome. Here we focus on the phenotype of Rlf null mutant mice generated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service