Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1411667

Sigma-Aldrich

Anti-MAGED1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 86.2 kDa

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAGED1(9500)

General description

This gene is a member of the melanoma antigen gene (MAGE) family. Most of the genes of this family encode tumor specific antigens that are not expressed in normal adult tissues except testis. Although the protein encoded by this gene shares strong homology with members of the MAGE family, it is expressed in almost all normal adult tissues. This gene has been demonstrated to be involved in the p75 neurotrophin receptor mediated programmed cell death pathway. Three transcript variants encoding two different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

MAGED1 (NP_001005332.1, 1 a.a. ~ 778 a.a) full-length human protein.

Sequence
MAQKMDCGAGLLGFQAEASVEDSALLMQTLMEAIQISEAPPTNQATAAASPQSSQPPTANEMADIQVSAAAARPKSAFKVQNATTKGPNGVYDFSQAHNAKDVPNTQPKAAFKSQNATPKGPNAAYDFSQAATTGELAANKSEMAFKAQNATTKVGPNATYNFSQSLNANDLANSRPKTPFKAWNDTTKAPTADTQTQNVNQAKMATSQADIETDPGISEPDGATAQTSADGSQAQNLESRTIIRGKRTRKINNLNVEENSSGDQRRAPLAAGTWRSAPVPVTTQNPPGAPPNVLWQTPLAWQNPSGWQNQTARQTPPARQSPPARQTPPAWQNPVAWQNPVIWPNPVIWQNPVIWPNPIVWPGPVVWPNPLAWQNPPGWQTPPGWQTPPGWQGPPDWQGPPDWPLPPDWPLPPDWPLPTDWPLPPDWIPADWPIPPDWQNLRPSPNLRPSPNSRASQNPGAAQPRDVALLQERANKLVKYLMLKDYTKVPIKRSEMLRDIIREYTDVYPEIIERACFVLEKKFGIQLKEIDKEEHLYILISTPESLAGILGTTKDTPKLGLLLVILGVIFMNGNRASEAVLWEALRKMGLRPGVRHPLLGDLRKLLTYEFVKQKYLDYRRVPNSNPPEYEFLWGLRSYHETSKMKVLRFIAEVQKRDPRDWTAQFMEAADEALDALDAAAAEAEARAEARTRMGIGDEAVSGPWSWDDIEFELLTWDEEGDFGDPWSRIPFTFWARYHQNARSRFPQTFAGPIIGPGGTASANFAANFGAIGFFWVE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service