Skip to Content
MilliporeSigma
All Photos(4)

Documents

SAB1411033

Sigma-Aldrich

Anti-DARS2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ASPRS, FLJ10514, LBSL, MT-ASPRS, RP3-383J4.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 73.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DARS2(55157)

General description

Aspartyl-tRNA synthetase 2, mitochondrial (DARS2) is a homodimeric enzyme. The gene encoding it is localized on human chromosome 1q25.1.

Immunogen

DARS2 (NP_060592.2, 1 a.a. ~ 645 a.a) full-length human protein.

Sequence
MYFPSWLSQLYRGLSRPIRRTTQPIWGSLYRSLLQSSQRRIPEFSSFVVRTNTCGELRSSHLGQEVTLCGWIQYRRQNTFLVLRDFDGLVQVIIPQDESAASVKKILCEAPVESVVQVSGTVISRPAGQENPKMPTGEIEIKVKTAELLNACKKLPFEIKNFVKKTEALRLQYRYLDLRSFQMQYNLRLRSQMVMKMREYLCNLHGFVDIETPTLFKRTPGGAKEFLVPSREPGKFYSLPQSPQQFKQLLMVGGLDRYFQVARCYRDEGSRPDRQPEFTQIDIEMSFVDQTGIQSLIEGLLQYSWPNDKDPVVVPFPTMTFAEVLATYGTDKPDTRFGMKIIDISDVFRNTEIGFLQDALSKPHGTVKAICIPEGAKYLKRKDIESIRNFAADHFNQEILPVFLNANRNWNSPVANFIMESQRLELIRLMETQEEDVVLLTAGEHNKACSLLGKLRLECADLLETRGVVLRDPTLFSFLWVVDFPLFLPKEENPRELESAHHPFTAPHPSDIHLLYTEPKKARSQHYDLVLNGNEIGGGSIRIHNAELQRYILATLLKEDVKMLSHLLQALDYGAPPHGGIALGLDRLICLVTGSPSIRDVIAFPKSFRGHDLMSNTPDSVPPEELKPYHIRVSKPTDSKAERAH

Biochem/physiol Actions

Aspartyl-tRNA synthetase 2, mitochondrial (DARS2) has a crucial role in the attachment of aspartate to the right mitochondrial transfer RNA. Mutations in the gene encoding it have been associated with leukoencephalopathy with brainstem and spinal cord involvement and elevated lactate (LBSL).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel homozygous mutation of DARS2 may cause a severe LBSL variant.
Miyake N
Clinical Genetics, 80(3), 293-296 (2011)
Pathogenic mutations causing LBSL affect mitochondrial aspartyl-tRNA synthetase in diverse ways.
van Berge L
The Biochemical Journal, 450(2), 345-350 (2013)
DARS-associated leukoencephalopathy can mimic a steroid-responsive neuroinflammatory disorder.
Wolf NI, et.l
Neurology, 84(3), 226-230 (2015)
Laura van Berge et al.
Brain : a journal of neurology, 137(Pt 4), 1019-1029 (2014-02-26)
Leukoencephalopathy with brainstem and spinal cord involvement and lactate elevation is a disorder caused by recessive mutations in the gene DARS2, which encodes mitochondrial aspartyl-tRNA synthetase. Recent observations indicate that the phenotypic range of the disease is much wider than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service