Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1410952

Sigma-Aldrich

Anti-COQ3 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

UG0215E05, bA9819.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 41 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COQ3(51805)

Related Categories

General description

Ubiquinone, also known as coenzyme Q, or Q, is a critical component of the electron transport pathways of both eukaryotes and prokaryotes (Jonassen and Clarke, 2000 [PubMed 10777520]). This lipid consists of a hydrophobic isoprenoid tail and a quinone head group. The tail varies in length depending on the organism, but its purpose is to anchor coenzyme Q to the membrane. The quinone head group is responsible for the activity of coenzyme Q in the respiratory chain. The S. cerevisiae COQ3 gene encodes an O-methyltransferase required for 2 steps in the biosynthetic pathway of coenzyme Q. This enzyme methylates an early coenzyme Q intermediate, 3,4-dihydroxy-5-polyprenylbenzoic acid, as well as the final intermediate in the pathway, converting demethyl-ubiquinone to coenzyme Q. The COQ3 gene product is also capable of methylating the distinct prokaryotic early intermediate 2-hydroxy-6-polyprenyl phenol.[supplied by OMIM

Immunogen

COQ3 (AAH63463.1, 1 a.a. ~ 369 a.a) full-length human protein.

Sequence
MWSGRKLGSSGGWFLRVLGPGGCNTKAARPLISSAVYVKNQLSGTLQIKPGVFNEYRTIWFKSYRTIFSCLNRIKSFRYPWARLYSTSQTTVDSGEVKTFLALAHKWWDEQGVYAPLHSMNDLRVPFIRDNLLKTIPNHQPGKPLLGMKILDVGCGGGLLTEPLGRLGASVIGIDPVDENIKTAQCHKSFDPVLDKRIEYRVCSLEEIVEETAETFDAVVASEVVEHVIDLETFLQCCCQVLKPGGSLFITTINKTQLSYALGIVFSEQIAGIVPKGTHTWEKFVSPETLESILESNGLSVQTVVGMLYNPFSGYWHWSENTSLNYAAHAVKSRVQEHPASAEFVLKGETEELQANACTNPAVHEKLKK

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service