Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1410493

Sigma-Aldrich

Anti-CNTNAP2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

AUTS15, CASPR2, CDFE, DKFZp781D1846, NRXN4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 148.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CNTNAP2(26047)

Related Categories

General description

This gene encodes a member of the neurexin family which functions in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, thrombospondin N-terminal-like domains and a putative PDZ binding site. This protein is localized at the juxtaparanodes of myelinated axons and associated with potassium channels. It may play a role in the local differentiation of the axon into distinct functional subdomains. This gene encompasses almost 1.5% of chromosome 7 and is one of the largest genes in the human genome. It may represent a positional candidate gene for the DFNB13 form of nonsyndromic deafness. (provided by RefSeq)

Immunogen

CNTNAP2 (NP_054860.1, 1 a.a. ~ 1331 a.a) full-length human protein.

Sequence
MQAAPRAGCGAALLLWIVSSCLCRAWTAPSTSQKCDEPLVSGLPHVAFSSSSSISGSYSPGYAKINKRGGAGGWSPSDSDHYQWLQVDFGNRKQISAIATQGRYSSSDWVTQYRMLYSDTGRNWKPYHQDGNIWAFPGNINSDGVVRHELQHPIIARYVRIVPLDWNGEGRIGLRIEVYGCSYWADVINFDGHVVLPYRFRNKKMKTLKDVIALNFKTSESEGVILHGEGQQGDYITLELKKAKLVLSLNLGSNQLGPIYGHTSVMTGSLLDDHHWHSVVIERQGRSINLTLDRSMQHFRTNGEFDYLDLDYEITFGGIPFSGKPSSSSRKNFKGCMESINYNGVNITDLARRKKLEPSNVGNLSFSCVEPYTVPVFFNATSYLEVPGRLNQDLFSVSFQFRTWNPNGLLVFSHFADNLGNVEIDLTESKVGVHINITQTKMSQIDISSGSGLNDGQWHEVRFLAKENFAILTIDGDEASAVRTNSPLQVKTGEKYFFGGFLNQMNNSSHSVLQPSFQGCMQLIQVDDQLVNLYEVAQRKPGSFANVSIDMCAIIDRCVPNHCEHGGKCSQTWDSFKCTCDETGYSGATCHNSIYEPSCEAYKHLGQTSNYYWIDPDGSGPLGPLKVYCNMTEDKVWTIVSHDLQMQTPVVGYNPEKYSVTQLVYSASMDQISAITDSAEYCEQYVSYFCKMSRLLNTPDGSPYTWWVGKANEKHYYWGGSGPGIQKCACGIERNCTDPKYYCNCDADYKQWRKDAGFLSYKDHLPVSQVVVGDTDRQGSEAKLSVGPLRCQGDRNYWNAASFPNPSSYLHFSTFQGETSADISFYFKTLTPWGVFLENMGKEDFIKLELKSATEVSFSFDVGNGPVEIVVRSPTPLNDDQWHRVTAERNVKQASLQVDRLPQQIRKAPTEGHTRLELYSQLFVGGAGGQQGFLGCIRSLRMNGVTLDLEERAKVTSGFISGCSGHCTSYGTNCENGGKCLERYHGYSCDCSNTAYDGTFCNKDVGAFFEEGMWLRYNFQAPATNARDSSSRVDNAPDQQNSHPDLAQEEIRFSFSTTKAPCILLYISSFTTDFLAVLVKPTGSLQIRYNLGGTREPYNIDVDHRNMANGQPHSVNITRHEKTIFLKLDHYPSVSYHLPSSSDTLFNSPKSLFLGKVIETGKIDQEIHKYNTPGFTGCLSRVQFNQIAPLKAALRQTNASAHVHIQGELVESNCGASPLTLSPMSSATDPWHLDHLDSASADFPYNPGQGQAIRNGVNRNSAIIGGVIAVVIFTILCTLVFLIRYMFRHKGTYHTNEAKGAESAESADAAIMNNDPNFTETIDESKKEWLI

Biochem/physiol Actions

Contactin associated protein-like 2 (CNTNAP2) controls the spatial clustering of voltage-gated potassium channels Kv1.1 and Kv1.2. It has been reported that alteration in the CNTNAP2 gene expression causes mental retardation, autistic behavior, seizures and microcephaly.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Clemm von Hohenberg et al.
Journal of psychiatric research, 47(10), 1349-1356 (2013-07-23)
CNTNAP2 is a gene on chromosome 7 that has shown associations with autism and schizophrenia, and there is evidence that it plays an important role for neuronal synchronization and brain connectivity. In this study, we assessed the relationship between Diffusion
Srirangan Sampath et al.
PloS one, 8(10), e77906-e77906 (2013-10-23)
Multiple lines of genetic evidence suggest a role for CNTNAP2 in autism. To assess its population impact we studied 2148 common single nucleotide polymorphisms (SNPs) using transmission disequilibrium test (TDT) across the entire ~3.3 Mb CNTNAP2 locus in 186 (408

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service