Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1409675

Sigma-Aldrich

Monoclonal Anti-BMP7 antibody produced in mouse

clone 1H3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

OP-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$704.00

$704.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$704.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$704.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H3, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.41 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP7(655)

General description

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, the BMP encoded by this gene has a proposed role in early development and possible bone inductive activity. (provided by RefSeq)

Immunogen

BMP7 (NP_001710, 293 a.a. ~ 390 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
STGSKQRSQNRSKTPKNQEALRMANVAENSSSDQRQACKKHELYVSFRDLGWQDWIIAPEGYAAYYCEGECAFPLNSYMNATNHAIVQTLVHFINPET

Biochem/physiol Actions

As implied by their name, bone morphogenetic proteins (BMPs) promote and regulate bone development and growth. BMP-7 induces apoptosis in B-cells. It has a role in organ development and may have a role in liver regeneration. Mutations in the gene encoding the protein have been shown to affect the development of teeth, palate and other orofacial complexes.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Are bone morphogenetic protein-7 (BMP-7) serum levels correlated with development of hepatic fibrosis?
Aktug Demir N
Journal of Infection in Developing Countries, 8(5), 605-610 (2014)
BMP7 expression correlates with secondary drug resistance in mantle cell lymphoma.
Camara-Clayette V
PLoS ONE, 8(9), e73993-e73993 (2013)
BMP7 Gene involved in nonsyndromic orofacial clefts in Western Han Chinese.
Yu Q
Medicina Oral, patologia Oral y cirugia Bucal, 20(3), e298-e304 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service