Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1409482

Sigma-Aldrich

Anti-PIWIL2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ10351, HILI, MGC133049, PIWIL1L, mili

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 157.03 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIWIL2(55124)

General description

PIWIL2 belongs to the Argonaute family of proteins, which function in development and maintenance of germline stem cells (Sasaki et al., 2003 [PubMed 12906857]).[supplied by OMIM

Immunogen

PIWIL2 (NP_060538.2, 1 a.a. ~ 973 a.a) full-length human protein.

Sequence
MDPFRPSFRGQSPIHPSQCQAVRMPGCWPQASKPLDPALGRGAPAGRGHVFGKPEEPSTQRGPAQRESVGLVSMFRGLGIETVSKTPLKREMLPSGRGILGRGLSANLVRKDREELSPTFWDPKVLAAGDSKMAETSVGWSRTLGRGSSDASLLPLGRAAGGISREVDKPPCTFSTPSRGPPQLSSPPALPQSPLHSPDRPLVLTVEHKEKELIVKQGSKGTPQSLGLNLVKIQCHNEAVYQYHVTFSPNVECKSMRFGMLKDHQAVTGNVTAFDGSILYLPVKLQQVLELKSQRKTDSAEISIKIQMTKILEPCSDLCIPFYNVVFRRVMKLLDMKLVGRNFYDPTSAMVLQQHRLQIWPGYAASIRRTDGGLFLLADVSHKVIRNDCVLDVMHAIYQQNKEHFQDECTKLLVGNIVITRYNNRTYRIDDVDWNKTPKDSFTMSDGKEITFLEYYSKNYGITVKEEDQPLLIHRPSERQDNHGMLLKGEILLLPELSFMTGIPEKMKKDFRAMKDLAQQINLSPKQHHSALECLLQRIAKNEAATNELMRWGLRLQKDVHKIEGRVLPMERINLKNTSFITSQELNWVKEVTRDPSILTIPMHFWALFYPKRAMDQARELVNMLEKIAGPIGMRMSPPAWVELKDDRIETYVRTIQSTLGAEGKIQMVVCIIMGPRDDLYGAIKKLCCVQSPVPSQVVNVRTIGQPTRLRSVAQKILLQINCKLGGELWGVDIPLKQLMVIGMDVYHDPSRGMRSVVGFVASINLTLTKWYSRVVFQMPHQEIVDSLKLCLVGSLKKFYEVNHCLPEKIVVYRDGVSDGQLKTVANYEIPQLQKCFEAFENYQPKMVVFVVQKKISTNLYLAAPQNFVTPTPGTVVDHTITSCEWVDFYLLAHHVRQGCGIPTHYVCVLNTANLSPDHMQRLTFKLCHMYWNWPGTIRVPAPCKYAHKLAFLSGHILHHEPAIQLCENLFFL

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service