Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1409291

Sigma-Aldrich

Monoclonal Anti-CUEDC1 antibody produced in mouse

clone 4F9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp547L163, FLJ20739

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$704.00

$704.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$704.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$704.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F9, monoclonal

form

buffered aqueous solution

mol wt

antigen 68.7 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUEDC1(404093)

Immunogen

CUEDC1 (NP_060419.1, 1 a.a. ~ 386 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MTSLFRRSSSGSGGGGTAGARGGGGGTAAPQELNNSRPARQVRRLEFNQAMDDFKTMFPNMDYDIIECVLRANSGAVDATIDQLLQMNLEGGGSSGGVYEDSSDSEDSIPPEILERTLEPDSSDEEPPPVYSPPAYHMHVFDRPYPLAPPTPPPRIDALGSGAPTSQRRYRNWNPPLLGNLPDDFLRILPQQLDSIQGNAGGPKPGSGEGCPPAMAGPGPGDQESRWKQYLEDERIALFLQNEEFMKELQRNRDFLLALERDRLKYESQKSKSSSVAVGNDFGFSSPVPGTGDANPAVSEDALFRDKLKHMGKSTRRKLFELARAFSEKTKMRKSKRKHLLKHQSLGAAASTANLLDDVEGHACDEDFRGRRQEAPKVEEGLREGQ

Biochem/physiol Actions

CUEDC1 (CUE domain containing 1) might be involved in protein trafficking and degradation. Changes in this gene are linked with early stage cervical cancer. It is highly methylated in the placental tissue of preeclampsia patients. CUEDC1 is suggested to control trophoblast invasion by participating in the TGF (transforming growth factor)-β/Smad (mothers against decapentaplegic homolog) signaling pathway. The gene is mapped to human chromosome 17q23.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y-H Yan et al.
European review for medical and pharmacological sciences, 17(22), 3083-3094 (2013-12-05)
Preeclampsia is a complication of pregnancy that severely threatens the health of the mother and infant, yet the mechanism of pathogenesis remains unclear. In this article, gene array technology was applied to identify the genes related to the pathogenesis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service