Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1409279

Sigma-Aldrich

Anti-LAMB1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CLM, MGC142015

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 198.1 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMB1(3912)

General description

Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Laminins are composed of 3 non identical chains: laminin alpha, beta and gamma (formerly A, B1, and B2, respectively) and they form a cruciform structure consisting of 3 short arms, each formed by a different chain, and a long arm composed of all 3 chains. Each laminin chain is a multidomain protein encoded by a distinct gene. Several isoforms of each chain have been described. Different alpha, beta and gamma chain isomers combine to give rise to different heterotrimeric laminin isoforms which are designated by Arabic numerals in the order of their discovery, i.e. alpha1beta1gamma1 heterotrimer is laminin 1. The biological functions of the different chains and trimer molecules are largely unknown, but some of the chains have been shown to differ with respect to their tissue distribution, presumably reflecting diverse functions in vivo. This gene encodes the beta chain isoform laminin, beta 1. The beta 1 chain has 7 structurally distinct domains which it shares with other beta chain isomers. The C-terminal helical region containing domains I and II are separated by domain alpha, domains III and V contain several EGF-like repeats, and domains IV and VI have a globular conformation. Laminin, beta 1 is expressed in most tissues that produce basement membranes, and is one of the 3 chains constituting laminin 1, the first laminin isolated from Engelbreth-Holm-Swarm (EHS) tumor. A sequence in the beta 1 chain that is involved in cell attachment, chemotaxis, and binding to the laminin receptor was identified and shown to have the capacity to inhibit metastasis. (provided by RefSeq)

Immunogen

LAMB1 (NP_002282.1, 1 a.a. ~ 1786 a.a) full-length human protein.

Sequence
MGLLQLLAFSFLALCRARVRAQEPEFSYGCAEGSCYPATGDLLIGRAQKLSVTSTCGLHKPEPYCIVSHLQEDKKCFICNSQDPYHETLNPDSHLIENVVTTFAPNRLKIWWQSENGVENVTIQLDLEAEFHFTHLIMTFKTFRPAAMLIERSSDFGKTWGVYRYFAYDCEASFPGISTGPMKKVDDIICDSRYSDIEPSTEGEVIFRALDPAFKIEDPYSPRIQNLLKITNLRIKFVKLHTLGDNLLDSRMEIREKYYYAVYDMVVRGNCFCYGHASECAPVDGFNEEVEGMVHGHCMCRHNTKGLNCELCMDFYHDLPWRPAEGRNSNACKKCNCNEHSISCHFDMAVYLATGNVSGGVCDDCQHNTMGRNCEQCKPFYYQHPERDIRDPNFCERCTCDPAGSQNEGICDSYTDFSTGLIAGQCRCKLNVEGEHCDVCKEGFYDLSSEDPFGCKSCACNPLGTIPGGNPCDSETGHCYCKRLVTGQHCDQCLPEHWGLSNDLDGCRPCDCDLGGALNNSCFAESGQCSCRPHMIGRQCNEVEPGYYFATLDHYLYEAEEANLGPGVSIVERQYIQDRIPSWTGAGFVRVPEGAYLEFFIDNIPYSMEYDILIRYEPQLPDHWEKAVITVQRPGRIPTSSRCGNTIPDDDNQVVSLSPGSRYVVLPRPVCFEKGTNYTVRLELPQYTSSDSDVESPYTLIDSLVLMPYCKSLDIFTVGGSGDGVVTNSAWETFQRYRCLENSRSVVKTPMTDVCRNIIFSISALLHQTGLACECDPQGSLSSVCDPNGGQCQCRPNVVGRTCNRCAPGTFGFGPSGCKPCECHLQGSVNAFCNPVTGQCHCFQGVYARQCDRCLPGHWGFPSCQPCQCNGHADDCDPVTGECLNCQDYTMGHNCERCLAGYYGDPIIGSGDHCRPCPCPDGPDSGRQFARSCYQDPVTLQLACVCDPGYIGSRCDDCASGYFGNPSEVGGSCQPCQCHNNIDTTDPEACDKETGRCLKCLYHTEGEHCQFCRFGYYGDALRQDCRKCVCNYLGTVQEHCNGSDCQCDKATGQCLCLPNVIGQNCDRCAPNTWQLASGTGCDPCNCNAAHSFGPSCNEFTGQCQCMPGFGGRTCSECQELFWGDPDVECRACDCDPRGIETPQCDQSTGQCVCVEGVEGPRCDKCTRGYSGVFPDCTPCHQCFALWDVIIAELTNRTHRFLEKAKALKISGVIGPYRETVDSVERKVSEIKDILAQSPAAEPLKNIGNLFEEAEKLIKDVTEMMAQVEVKLSDTTSQSNSTAKELDSLQTEAESLDNTVKELAEQLEFIKNSDIRGALDSITKYFQMSLEAEERVNASTTEPNSTVEQSALMRDRVEDVMMERESQFKEKQEEQARLLDELAGKLQSLDLSAAAEMTCGTPPGASCSETECGGPNCRTDEGERKCGGPGCGGLVTVAHNAWQKAMDLDQDVLSALAEVEQLSKMVSEAKLRADEAKQSAEDILLKTNATKEKMDKSNEELRNLIKQIRNFLTQDSADLDSIEAVANEVLKMEMPSTPQQLQNLTEDIRERVESLSQVEVILQHSAADIARAEMLLEEAKRASKSATDVKVTADMVKEALEEAEKAQVAAEKAIKQADEDIQGTQNLLTSIESETAASEETLFNASQRISELERNVEELKRKAAQNSGEAEYIEKVVYTVKQSAEDVKKTLDGELDEKYKKVENLIAKKTEESADARRKAEMLQNEAKTLLAQANSKLQLLKDLERKYEDNQRYLEDKAQELARLEGEVRSLLKDISQKVAVYSTCL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service