Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1409066

Sigma-Aldrich

Monoclonal Anti-GCA antibody produced in mouse

clone 3D12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

GCL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
clone:
3D12, monoclonal
species reactivity:
human
citations:
technique(s):
indirect ELISA: suitable

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D12, monoclonal

form

buffered aqueous solution

mol wt

antigen 49.61 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCA(25801)

General description

This gene product, grancalcin, is a calcium-binding protein abundant in neutrophils and macrophages. It belongs to the penta-EF-hand subfamily of proteins which includes sorcin, calpain, and ALG-2. Grancalcin localization is dependent upon calcium and magnesium. In the absence of divalent cation, grancalcin localizes to the cytosolic fraction; with magnesium alone, it partitions with the granule fraction; and in the presence of magnesium and calcium, it associates with both the granule and membrane fractions, suggesting a role for grancalcin in granule-membrane fusion and degranulation. (provided by RefSeq)

Immunogen

GCA (AAH05214, 1 a.a. ~ 217 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAYPGYGGGFGNFSIQVPGMQMGQPVPETGPAILLDGYSGPAYSDTYSSAGDSVYTYFSAVAGQDGEVDAEELQRCLTQSGINGTYSPFSLETCRIMIAMLDRDHTGKMGFNAFKELWAALNAWKENFMTVDQDGSGTVEHHELRQAIGLMGYRLSPQTLTTIVKRYSKNGRIFFDDYVACCVKLRALTDFFRKRDHLQQGSANFIYDDFLQGTMAI

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service