Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1407456

Sigma-Aldrich

Anti-CRBN antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CRBN Antibody - Anti-CRBN antibody produced in mouse, Cereblon Antibody, Crbn Antibody, DKFZp781K0715, MGC27358, MRT2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50.5 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRBN(51185)

General description

Cereblon (CRBN) is encoded by the gene mapped to human chromosome 3p26.2. It is ubiquitously expressed in various subcellular compartments such as nucleus, cytoplasm and endoplasmic reticulum (ER).

Immunogen

CRBN (NP_057386.2, 1 a.a. ~ 442 a.a) full-length human protein.

Sequence
MAGEGDQQDAAHNMGNHLPLLPAESEEEDEMEVEDQDSKEAKKPNIINFDTSLPTSHTYLGADMEEFHGRTLHDDDSCQVIPVLPQVMMILIPGQTLPLQLFHPQEVSMVRNLIQKDRTFAVLAYSNVQEREAQFGTTAEIYAYREEQDFGIEIVKVKAIGRQRFKVLELRTQSDGIQQAKVQILPECVLPSTMSAVQLESLNKCQIFPSKPVSREDQCSYKWWQKYQKRKFHCANLTSWPRWLYSLYDAETLMDRIKKQLREWDENLKDDSLPSNPIDFSYRVAACLPIDDVLRIQLLKIGSAIQRLRCELDIMNKCTSLCCKQCQETEITTKNEIFSLSLCGPMAAYVNPHGYVHETLTVYKACNLNLIGRPSTEHSWFPGYAWTVAQCKICASHIGWKFTATKKDMSPQKFWGLTRSALLPTIPDTEDEISPDKVILCL

Application

Anti-CRBN antibody produced in mouse has been used in western blotting.

Biochem/physiol Actions

Cereblon (CRBN) is a substrate binding component of the E3 ubiquitin ligase complex. It interacts with thalidomide and its analog to facilitate proteasome-dependent degradation of the transcription factors, Ikaros family zinc finger protein 1 and 3 (IKZF1 and IKZF3). CRBN provides cytoprotection against the ubiquitin-proteasome system-dysfunction. It negatively regulates the AMP-activated protein kinase signaling pathway in vivo. Nonsense mutation of the CRBN gene causes mild type of autosomal recessive nonsyndromic intellectual disability (ID).

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunomodulatory drugs inhibit TLR4-induced type-1 interferon production independently of Cereblon via suppression of the TRIF/IRF3 pathway
Millrine D, et al.
International Immunology, 28(6), 307-315 (2016)
Cereblon is a direct protein target for immunomodulatory and antiproliferative activities of lenalidomide and pomalidomide
Lopez-Girona AEA, et al.
Leukemia Lymphoma, 26(11), 2326-2335 (2012)
Mitochondrial cereblon functions as a Lon-type protease
Kataoka K, et al.
Scientific reports, 6, 29986-29986 (2016)
David Millrine et al.
International immunology, 28(6), 307-315 (2016-02-13)
Thalidomide and its derivatives, collectively referred to as immunomodulatory drugs (IMiDs), are effective inhibitors of inflammation and are known to inhibit TLR-induced TNFα production. The identification of Cereblon as the receptor for these compounds has led to a rapid advancement
Cereblon is recruited to aggresome and shows cytoprotective effect against ubiquitin-proteasome system dysfunction
Sawamura N, et al.
Biochemical and biophysical research communications, 464(4), 1054-1059 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service