Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB1407370

Sigma-Aldrich

Anti-IFT81 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CDV-1, CDV-1R, CDV1, CDV1R, MGC102777, MGC4027

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~79.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFT81(28981)

Related Categories

General description

Mouse polyclonal antibody raised against a full-length human IFT81 protein. Intraflagellar transport 81 (IFT81) is part of the intraflagellar transport (IFT) B-complex. The gene encoding it is localized on human chromosome 12q24.

Immunogen

IFT81 (NP_054774.2, 1 a.a. ~ 676 a.a) full-length human protein.

Sequence
MSDQIKFIMDSLNKEPFRKNYNLITFDSLEPMQLLQVLSDVLAEIDPKQLVDIREEMPEQTAKRMLSLLGILKYKPSGNATDMSTFRQGLVIGSKPVIYPVLHWLLQRTNELKKRAYLARFLIKLEVPSEFLQDETVADTNKQYEELMEAFKTLHKEYEQLKISGFSTAEIRKDISAMEEEKDQLIKRVEHLKKRVETAQNHQWMLKIARQLRVEKEREEYLAQQKQEQKNQLFHAVQRLQRVQNQLKSMRQAAADAKPESLMKRLEEEIKFNLYMVTEKFPKELENKKKELHFLQKVVSEPAMGHSDLLELESKINEINTEINQLIEKKMMRNEPIEGKLSLYRQQASIISRKKEAKAEELQEAKEKLASLEREASVKRNQTREFDGTEVLKGDEFKRYVNKLRSKSTVFKKKHQIIAELKAEFGLLQRTEELLKQRHENIQQQLQTMEEKKGISGYSYTQEELERVSALKSEVDEMKGRTLDDMSEMVKKLYSLVSEKKSALASVIKELRQLRQKYQELTQECDEKKSQYDSCAAGLESNRSKLEQEVRRLREECLQEESRYHYTNCMIKNLEVQLRRATDEMKAYISSDQQEKRKAIREQYTKNTAEQENLGKKLREKQKVIRESHGPNMKQAKMWRDLEQLMECKKQCFLKQQSQTSIGQVIQEGGEDRLIL

Biochem/physiol Actions

Intraflagellar transport 81 (IFT81) associates with intraflagellar transport 74 (IFT74) to form a complex which binds to tubulin. This has a role in ciliogenesis. Mutations in the gene encoding IFT81 have been linked to ciliopathy.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genomic organization and mapping of mouse CDV (carnitine deficiency-associated gene expressed in ventricle)-1 and its related CDV-1R gene.
Higashi M
Mammalian Genome, 11(12), 1053-1057 (2000)
Isabelle Perrault et al.
Journal of medical genetics, 52(10), 657-665 (2015-08-16)
Bidirectional intraflagellar transport (IFT) consists of two major protein complexes, IFT-A and IFT-B. In contrast to the IFT-B complex, all components of IFT-A have recently been linked to human ciliopathies when defective. We therefore hypothesised that mutations in additional IFT-B
Sagar Bhogaraju et al.
Science (New York, N.Y.), 341(6149), 1009-1012 (2013-08-31)
Intraflagellar transport (IFT) of ciliary precursors such as tubulin from the cytoplasm to the ciliary tip is involved in the construction of the cilium, a hairlike organelle found on most eukaryotic cells. However, the molecular mechanisms of IFT are poorly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service