Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1407313

Sigma-Aldrich

Anti-OR8B8 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

TPCR85

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$639.00

$639.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
$639.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$639.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~34.5 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OR8B8(26493)

General description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. (provided by RefSeq)

Immunogen

OR8B8 (NP_036510.1, 1 a.a. ~ 311 a.a) full-length human protein.

Sequence
MAAENSSFVTQFILAGLTDQPGVQIPLFFLFLGFYVVTVVGNLGLITLIRLNSHLHTPMYFFLYNLSFIDFCYSSVITPKMLMSFVLKKNSISYAGCMTQLFFFLFFVVSESFILSAMAYDRYVAICNPLLYMVTMSPQVCFLLLLGVYGMGFAGAMAHTACMMGVTFCANNLVNHYMCDILPLLECACTSTYVNELVVFVVVGIDIGVPTVTIFISYALILSSIFHIDSTEGRSKAFSTCSSHIIAVSLFFGSGAFMYLKPFSLLAMNQGKVSSLFYTTVVPMLNPLIYSLRNKDVKVALKKILNKNAFS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service