Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1407067

Sigma-Aldrich

Anti-ACTL7A antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~48.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTL7A(10881)

Related Categories

General description

The protein encoded by this gene is a member of a family of actin-related proteins (ARPs) which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene (ACTL7A), and related gene, ACTL7B, are intronless, and are located approximately 4 kb apart in a head-to-head orientation within the familial dysautonomia candidate region on 9q31. Based on mutational analysis of the ACTL7A gene in patients with this disorder, it was concluded that it is unlikely to be involved in the pathogenesis of dysautonomia. The ACTL7A gene is expressed in a wide variety of adult tissues, however, its exact function is not known. (provided by RefSeq)

Immunogen

ACTL7A (NP_006678.1, 1 a.a. ~ 435 a.a) full-length human protein.

Sequence
MWAPPAAIMGDGPTKKVGNQAPLQTQALQTASLRDGPAKRAVWVRHTSSEPQEPTESKAAKERPKQEVTKAVVVDLGTGYCKCGFAGLPRPTHKISTTVGKPYMETAKTGDNRKETFVGQELNNTNVHLKLVNPLRHGIIVDWDTVQDIWEYLFRQEMKIAPEEHAVLVSDPPLSPHTNREKYAEMLFEAFNTPAMHIAYQSRLSMYSYGRTSGLVVEVGHGVSYVVPIYEGYPLPSITGRLDYAGSDLTAYLLGLLNSAGNEFTQDQMGIVEDIKKKCCFVALDPIEEKKVPLSEHTIRYVLPDGKEIQLCQERFLCSEMFFKPSLIKSMQLGLHTQTVSCLNKCDIALKRDLMGNILLCGGSTMLSGFPNRLQKELSSMCPNDTPQVNVLPERDSAVWTGGSILASLQGFQPLWVHRFEYEEHGPFFLYRRCF

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service