Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB1406495

Sigma-Aldrich

Anti-AURKC antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

AIE2, AIK3, AurC, STK13, aurora-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~35.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AURKC(6795)

Related Categories

General description

This gene encodes a member of the Aurora subfamily of serine/threonine protein kinases. The encoded protein is a chromosomal passenger protein that forms complexes with Aurora-B and inner centromere proteins and may play a role in organizing microtubules in relation to centrosome/spindle function during mitosis. This gene is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction. Alternative splicing results in multiple transcript variants. (provided by RefSeq)

Immunogen

AURKC (NP_001015878.1, 1 a.a. ~ 309 a.a) full-length human protein.

Sequence
MSSPRAVVQLGKAQPAGEELATANQTAQQPSSPAMRRLTVDDFEIGRPLGKGKFGNVYLARLKESHFIVALKVLFKSQIEKEGLEHQLRREIEIQAHLQHPNILRLYNYFHDARRVYLILEYAPRGELYKELQKSEKLDEQRTATIIEELADALTYCHDKKVIHRDIKPENLLLGFRGEVKIADFGWSVHTPSLRRKTMCGTLDYLPPEMIEGRTYDEKVDLWCIGVLCYELLVGYPPFESASHSETYRRILKVDVRFPLSMPLGARDLISRLLRYQPLERLPLAQILKHPWVQAHSRRVLPPCAQMAS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service