Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1405647

Sigma-Aldrich

Anti-COX10 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~48.73 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COX10(1352)

General description

Cytochrome c oxidase (COX), the terminal component of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. This component is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may function in the regulation and assembly of the complex. This nuclear gene encodes heme A:farnesyltransferase, which is not a structural subunit but required for the expression of functional COX and functions in the maturation of the heme A prosthetic group of COX. This protein is predicted to contain 7-9 transmembrane domains localized in the mitochondrial inner membrane. A gene mutation, which results in the substitution of a lysine for an asparagine (N204K), is identified to be responsible for cytochrome c oxidase deficiency. In addition, this gene is disrupted in patients with CMT1A (Charcot-Marie-Tooth type 1A) duplication and with HNPP (hereditary neuropathy with liability to pressure palsies) deletion. (provided by RefSeq)

Immunogen

COX10 (AAH06394, 1 a.a. ~ 443 a.a) full-length human protein.

Sequence
MAASPHTLSSRLLTGCVGGSVWYLERRTIQDSPHKFLHLLRNVNKQWITFQHFSFLKRMYVTQLNRSHNQQVRPKPEPVASPFLEKTSSGQAKAEIYEMRPLSPPSLSLSRKPNEKELIELEPDSVIEDSIDVGKETKEEKRWKEMKLQVYDLPGILAQLSKIKLTALVVSTTAAGFALAPGPFDWPCFLLTSVGTGLASCAANSINQFFEVPFDSNMNRTKNRPLVRGQISPLLAVSFATCCAVPGVAILTLGVNPLTGALGLFNIFLYTCCYTPLKRISIANTWVGAVVGAIPPVMGWTAATGSLDAGAFLLGGILYSWQFPHFNALSWGLREDYSRGGYCMMSVTHPGLCRRVALRHCLALLVLSAAAPVLDITTWTFPIMALPINAYISYLGFRFYVDADRRSSRRLFFCSLWHLPLLLLLMLTCKRPSGGGDAGPPPS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service