Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB1405070

Sigma-Aldrich

Monoclonal Anti-CHST11 antibody produced in mouse

clone 1H3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

C4ST, C4ST-1, C4ST1, HSA269537

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.99 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CHST11(50515)

General description

Chondroitin 4-sulfotransferases, such as CHST11, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate to position 4 of N-acetylgalactosamine residues in chondroitin (Yamauchi et al., 2000 [PubMed 10722746]).[supplied by OMIM

Immunogen

CHST11 (NP_060883, 230 a.a. ~ 337 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
RKGDDVKFEEFVAYLIDPHTQREEPFNEHWQTVYSLCHPCHIHYDLVGKYETLEEDSNYVLQLAGVGSYLKFPTYAKSTRTTDEMTTEFFQNISSEHQTQLYEVYKLD

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasufumi Matsuda et al.
Cancer science, 107(5), 619-628 (2016-02-20)
An increasingly accepted concept is that the progression of colorectal cancer is accompanied by epithelial-mesenchymal transition (EMT). In our study, in order to characterize the properties of EMT in 16 colorectal cancer cell lines, the cells were first orthotopically implanted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service