Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB1404642

Sigma-Aldrich

Monoclonal Anti-SYNJ1 antibody produced in mouse

clone 1A2, ascites fluid

Synonym(s):

INPP5G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1A2, monoclonal

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYNJ1(8867)

General description

Synaptojanin 1 is a phosphoinositide phosphatase that regulates levels of membrane phosphatidylinositol-4,5-bisphosphate. As such, expression of this enzyme may affect synaptic transmission and membrane trafficking.[supplied by RefSeq

Immunogen

SYNJ1 (NP_003886, 1474 a.a. ~ 1573 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KISNPKGWVTFEEEEDFGVKGKSKSACSDLLGNQPSSFSGSNLTLNDDWNKGTNVSFCVLPSRRPPPPPVPLLPPGTSPPVDPFTTLASKASPTLDFTER

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service