Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1404299

Sigma-Aldrich

Monoclonal Anti-RARS antibody produced in mouse

clone 1E9-2B5, ascites fluid

Synonym(s):

ArgRS, DALRD1, MGC8641

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1E9-2B5, monoclonal

mol wt

antigen ~98.71 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RARS(5917)

General description

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Arginyl-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family. (provided by RefSeq)

Immunogen

RARS (AAH00528, 1 a.a. ~ 660 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDVLVSECSARLLQQEEEIKSLTAEIDRLKNCGCLGASPNLEQLQEENLKLKYRLNILRKSLQAERNKPTKNMINIISRLQEVFGHAIKAAYPDLENPPLLVTPSQQAKFGDYQCNSAMGISQMLKTKEQKVNPREIAENITKHLPDNECIEKVEIAGPGFINVHLRKDFVSEQLTSLLVNGVQLPALGENKKVIVDFSSPNIAKEMHVGHLRSTIIGESISRLFEFAGYDVLRLNHVGDWGTQFGMLIAHLQDKFPDYLTVSPPIGDLQVFYKESKKRFDTEEEFKKRAYQCVVLLQGKNPDITKAWKLICDVSRQELNKIYDALDVSLIERGESFYQDRMNDIVKEFEDRGFVQVDDGRKIVFVPGCSIPLTIVKSDGGYTYDTSDLAAIKQRLFEEKADMIIYVVDNGQSVHFQTIFAAAQMIGWYDPKVTRVFHAGFGVVLGEDKKKFKTRSGETVRLMDLLGEGLKRSMDKLKEKERDKVLTAEELNAAQTSVAYGCIKYADLSHNRLNDYIFSFDKMLDDRGNTAAYLLYAFTRIRSIARLANIDEEMLQKAARETKILLDHEKEWKLGRCILRFPEILQKILDDLFLHTLCDYIYELATAFTEFYDSCYCVEKDRQTGKILKVNMWRMLLCEAVAAVMAKGFDILGIKPVQRM

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service