Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1403429

Sigma-Aldrich

Monoclonal Anti-ACRC, (C-terminal) antibody produced in mouse

clone 2E4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

NAAR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACRC(93953)

General description

The gene ACRC (acidic repeat containing) is mapped to human chromosome Xq13.1 and contains at least 12 exons and 11 introns. It contains a characteristic acidic repeat region of 21 units of 8–10 amino acids. It is expressed ubiquitously, including skeletal muscle, liver, kidney, pancreas, heart, lung, and brain, with the highest expression in skeletal muscle.

Immunogen

ACRC (NP_443189.1, 592 a.a. ~ 691 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
HEMCHAASWLIDGIHDSHGDAWKYYARKSNRIHPELPRVTRCHNYKINYKVHYECTGCKTRIGCYTKSLDTSRFICAKCKGSLVMVPLTQKDGTRIVPHV

Biochem/physiol Actions

The gene ACRC (acidic repeat containing) is localized on a critical region that harbors the disease locus for X-linked dystonia parkinsonism (XDP). However, mutation analysis in this gene does not implicate it in the disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ACRC codes for a novel nuclear protein with unusual acidic repeat tract and maps to DYT3 (dystonia parkinsonism) critical interval in xq13.1.
Nolte D
Neurogenetics, 3, 207-213 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service