Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB1402916

Sigma-Aldrich

Monoclonal Anti-SIPA1, (C-terminal) antibody produced in mouse

clone 2B4, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2B4, monoclonal

mol wt

antigen ~38.1 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIPA1(6494)

Related Categories

General description

The product of this gene is a mitogen induced GTPase activating protein (GAP). It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date. (provided by RefSeq)

Immunogen

SIPA1 (AAH10492, 933 a.a. ~ 1042 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SEIASTCNTILESLSREGQPIPESGDPKGTPKSDAEPEPGNLSEKVSHLESMLRKLQEDLQKEKADRAALEEEVRSLRHNNRRLQAESESAATRLLLASKQLGSPTADLA

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service