This product has not been tested for use in flow cytometry. There are no current anti-TSPAN2 products tested for use in flow.
SAB1402499
Monoclonal Anti-TSPAN2 antibody produced in mouse
clone 4A3, purified immunoglobulin, buffered aqueous solution
Synonym(s):
6330415F13Rik, FLJ12082, TSN2, TSPAN-2
Select a Size
$715.00
Select a Size
About This Item
$715.00
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
4A3, monoclonal
form
buffered aqueous solution
mol wt
antigen ~34.47 kDa
species reactivity
human
technique(s)
capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL
isotype
IgG2a
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TSPAN2(10100)
General description
Immunogen
Sequence
GKGVAIRHVQTMYEEAYNDYLKDRGKGNGTLITFHSTFQCCGKESSEQVQPTCPKELLGHKNCIDEIETIISVKLQ
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
does this antibody work for flow?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service