Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1402405

Sigma-Aldrich

Monoclonal Anti-ZNF165 antibody produced in mouse

clone 2B8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

LD65, ZSCAN7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~79.46 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF165(7718)

General description

This gene encodes a member of the Kruppel family of zinc finger proteins. Members of this DNA-binding protein family act as transcriptional regulators. This gene is located within a cluster of zinc finger family members. The encoded protein may play a role in spermatogenesis. (provided by RefSeq)

Immunogen

ZNF165 (AAH26092, 1 a.a. ~ 485 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MATEPKKAAAQNSPEDEGLLIVKIEEEEFIHGQDTCLQRSELLKQELCRQLFRQFCYQDSPGPREALSRLRELCCQWLKPEIHTKEQILELLVLEQFLTILPGDLQAWVHEHYPESGEEAVTILEDLERGTDEAVLQVQAHEHGQEIFQKKVSPPGPALNVKLQPVETKAHFDSSEPQLLWDCDNESENSRSMPKLEIFEKIESQRIISGRISGYISEASGESQDICKSAGRVKRQWEKESGESQRLSSAQDEGFGKILTHKNTVRGEIISHDGCERRLNLNSNEFTHQKSCKHGTCDQSFKWNSDFINHQIIYAGEKNHQYGKSFKSPKLAKHAAVFSGDKTHQCNECGKAFRHSSKLARHQRIHTGERCYECNECGKSFAESSDLTRHRRIHTGERPFGCKECGRAFNLNSHLIRHQRIHTREKPYECSECGKTFRVSSHLIRHFRIHTGEKPYECSECGRAFSQSSNLSQHQRIHMRENLLM

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service