Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1401851

Sigma-Aldrich

Anti-PNPLA3 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ADPN, C22orf20, iPLA(2)epsilon

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$754.00

$754.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$754.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$754.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PNPLA3(80339)

General description

Patatin like phospholipase domain containing 3 (PNPLA3) gene is mapped to human chromosome 22q13.3. The protein encoded by this gene is a triacylglycerol lipase.

Immunogen

PNPLA3 (AAH65195.1, 1 a.a. ~ 481 a.a) full-length human protein.

Sequence
MYDAERGWSLSFAGCGFLGFYHVGATRCLSEHAPHLLRDARMLFGASAGALHCVGVLSGIPLEQTLQVLSDLVRKARSRNIGIFHPSFNLSKFLRQGLGKCLPANVHQLISGKIGISLTRVSDGENVLVSDFRSKDEVVDALVCSCFMPFYSGLIPPSFRGVRYVDGGVSDNVPFIDAKTTITVSPFYGEYDICPKVKSTNFLHVDITKLSLRLCTGNLYLLSRAFVPPDLKVLGEICLRGYLDAFRFLEEKGICNRPQPGLKSSSEGMDPEVAMPSWANMSLDSSPESAALAVRLEGDELLDHLRLSILPWDESILDTLSPRLATALSEEMKDKGGYMSKICNLLPIRIMSYVMLPCTLPVESAIAIVQRLVTWLPDMPDDVLWLQWVTSQVFTRVLMCLLPASRSQMPVSSQQASPCTPEQDWPCWTPCSPEGCPAETKAEATPRSILRSSLNFFLGNKVPAGAEGLSTFPSFSLEKSL

Application

Anti-PNPLA3 antibody produced in rabbit has been used in western blotting.[1]

Biochem/physiol Actions

PNPLA3 (patatin like phospholipase domain containing 3) involves lipogenic transacetylase activity. Variation in PNPLA3 (patatin like phospholipase domain containing 3) is known to increase the fat content of hepatocytes, thereby promoting the severity of nonalcoholic liver disease. The gene is found to be associated with hepatic steatosis, liver fibrosis and cancer. PNPLA3 might be involved in energy homeostasis. PNPLA3 mediates triacylglycerol hydrolysis in adipocytes and is involved in the balance of energy usage cum storage in adipocytes.
PNPLA3 mediates triacylglycerol hydrolysis in adipocytes and is involved in the balance of energy usage cum storage in adipocytes.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The PNPLA3 I148M variant modulates the fibrogenic phenotype of human hepatic stellate cells.
Bruschi FV
Hepatology, 65(6), 1875-1890 (2017)
Variant in PNPLA3 is associated with alcoholic liver disease.
Tian C
Nature Genetics, 42(1), 21-23 (2010)
Cyanidin-3-O-beta-glucoside protects against liver fibrosis induced by alcohol via regulating energy homeostasis and AMPKautophagy signaling pathway
Wan T, et al.
Journal of functional foods, 37, 16-24 (2017)
Silvia Sookoian et al.
World journal of gastroenterology, 18(42), 6018-6026 (2012-11-17)
Genome-wide and candidate gene association studies have identified several variants that predispose individuals to developing nonalcoholic fatty liver disease (NAFLD). However, the gene that has been consistently involved in the genetic susceptibility of NAFLD in humans is patatin-like phospholipase domain
PNPLA3 and RNF7 Gene Variants are Associated with the Risk of Developing Liver Fibrosis and Cirrhosis in an Eastern European Population.
Kupcinskas J
Journal of Gastrointestinal and Liver Diseases : JGLD, 26(1), 37-43 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service