Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1401627

Sigma-Aldrich

Anti-SENP3 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp586K0919, DKFZp762A152, SMT3IP1, SSP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
technique(s):
western blot: 1 μg/mL

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SENP3(26168)

General description

The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins (see SUMO1; MIM 601912) is required for numerous biologic processes. SUMO-specific proteases, such as SENP3, are responsible for the initial processing of SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also have isopeptidase activity for the removal of SUMO from high molecular mass SUMO conjugates (Di Bacco et al., 2006 [PubMed 16738315]).[supplied by OMIM

Immunogen

SENP3 (NP_056485.2, 1 a.a. ~ 574 a.a) full-length human protein.

Sequence
MKETIQGTGSWGPEPPGPGIPPAYSSPRRERLRWPPPPKPRLKSGGGFGPDPGSGTTVPARRLPVPRPSFDASASEEEEEEEEEEDEDEEEEVAAWRLPPRWSQLGTSQRPRPSRPTHRKTCSQRRRRAMRAFRMLLYSKSTSLTFHWKLWGRHRGRRRGLAHPKNHLSPQQGGATPQVPSPCCRFDSPRGPPPPRLGLLGALMAEDGVRGSPPVPSGPPMEEDGLRWTPKSPLDPDSGLLSCTLPNGFGGQSGPEGERSLAPPDASILISNVCSIGDHVAQELFQGSDLGMAEEAERPGEKAGQHSPLREEHVTCVQSILDEFLQTYGSLIPLSTDEVVEKLEDIFQQEFSTPSRKGLVLQLIQSYQRMPGNAMVRGFRVAYKRHVLTMDDLGTLYGQNWLNDQVMNMYGDLVMDTVPEKVHFFNSFFYDKLRTKGYDGVKRWTKNVDIFNKELLLIPIHLEVHWSLISVDVRRRTITYFDSQRTLNRRCPKHIAKYLQAEAVKKDRLDFHQGWKGYFKMNVARQNNDSDCGAFVLQYCKHLALSQPFSFTQQDMPKLRRQIYKELCHCKLTV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service