Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1401563

Sigma-Aldrich

Anti-ZNF277 antibody produced in rabbit

whole antiserum, buffered aqueous solution

Synonym(s):

NRIF4, ZNF277P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF277(11179)

Immunogen

ZNF277 (ENSP00000355043, 1 a.a. ~ 438 a.a) full-length human protein.

Sequence
MQEDRDGSCSTVGGVGYGDSKDCILEPLSLPESPGGTTTLEGSPSVPCIFCEEHFPVAEQDKLLKHMIIEHKIVIADVKLVADFQRYILYWRKRFTEQPITDFCSVIRINSTAPFEEQENYFLLCDVLPEDRILREELQKQRLREILEQQQQERNDTNFHGVCMFCNEEFLGNRSVILNHMAREHAFNIGLPDNIVNCNEFLCTLQKKLDNLQCLYCEKTFRDKNTLKDHMRKKQHRKINPKNREYDRFYVINYLELGKSWEEVQLEDDRELLDHQEDDWSDWEEHPASAVCLFCEKQAETIEKLYVHMEDAHEFDLLKIKSELGLNFYQQVKLVNFIRRQVHQCRCYGCHVKFKSKADLRTHMEETKHTSLLPDRKTWDQLEYYFPTYENDTLLCTLSDSESDLTAQEQNENVPIISEDTSKLYALKQSSILNQLLL

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service