Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1401419

Sigma-Aldrich

Anti-MKKS antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

BBS6, HMCS, KMS, MKS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MKKS(8195)

General description

McKusick-Kaufman syndrome putative chaperonin (MKKS) is a centrosome-shuttling protein.The gene is located on human chromosome 20p12.2.It This protein encodes a chaperon-like protein.

Immunogen

MKKS (NP_061336.1, 1 a.a. ~ 570 a.a) full-length human protein.

Sequence
MSRLEAKKPSLCKSEPLTTERVRTTLSVLKRIVTSCYGPSGRLKQLHNGFGGYVCTTSQSSALLSHLLVTHPILKILTASIQNHVSSFSDCGLFTAILCCNLIENVQRLGLTPTTVIRLNKHLLSLCISYLKSETCGCRIPVDFSSTQILLCLVRSILTSKPACMLTRKETEHVSALILRAFLLTIPENAEGHIILGKSLIVPLKGQRVIDSTVLPGILIEMSEVQLMRLLPIKKSTALKVALFCTTLSGDTSDTGEGTVVVSYGVSLENAVLDQLLNLGRQLISDHVDLVLCQKVIHPSLKQFLNMHRIIAIDRIGVTLMEPLTKMTGTQPIGSLGSICPNSYGSVKDVCTAKFGSKHFFHLIPNEATICSLLLCNRNDTAWDELKLTCQTALHVLQLTLKEPWALLGGGCTETHLAAYIRHKTHNDPESILKDDECTQTELQLIAEAFCSALESVVGSLEHDGGEILTDMKYGHLWSVQADSPCVANWPDLLSQCGCGLYNSQEELNWSFLRSTRRPFVPQSCLPHEAVGSASNLTLDCLTAKLSGLQVAVETANLILDLSYVIEDKN

Biochem/physiol Actions

McKusick-Kaufman syndrome putative chaperonin (MKKS) plays a crucial role in cytokinesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MKKS is a centrosome-shuttling protein degraded by disease-causing mutations via CHIP-mediated ubiquitination
Hirayama , et al.
Molecular Biology of the Cell, 19(3), 899-911 (2008)
Molecular diagnosis reveals genetic heterogeneity for the overlapping MKKS and BBS phenotypes
Schaefer, et al.
European Journal of Medical Genetics, 54(2), 157-160 (2011)
MKKS/BBS6, a divergent chaperonin-like protein linked to the obesity disorder Bardet-Biedl syndrome, is a novel centrosomal component required for cytokinesis
Kim, Jun, et al.
Journal of Cell Science, 118(5), 1007-1020 (2005)
S P Daiger et al.
Clinical genetics, 84(2), 132-141 (2013-05-25)
Retinitis pigmentosa (RP) is a heterogeneous set of inherited retinopathies with many disease-causing genes, many known mutations, and highly varied clinical consequences. Progress in finding treatments is dependent on determining the genes and mutations causing these diseases, which includes both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service