Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1401272

Sigma-Aldrich

Monoclonal Anti-OMP antibody produced in mouse

clone 2B7, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OMP(4975)

Related Categories

General description

Olfactory marker protein is uniquely associated with the mature olfactory receptor neurons in many vertebrate species from fish to man. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Results of the mouse knockout studies show that OMP-null mice are compromised in their ability to respond to odor stimuli, and that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. (provided by RefSeq)

Immunogen

OMP (NP_006180.1, 65 a.a. ~ 163 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
FERWNVVLDKPGKVTITGTSQNWTPDLTNLMTRQLLDPTAIFWRKEDSDAIDWNEADALEFGERLSDLAKIRKVMYFLVTFGEGVEPANLKASVVFNQL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service