Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1400862

Sigma-Aldrich

Anti-C1QTNF9 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MGC48915

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

C1QTNF9 (AAH40438.1, 1 a.a. ~ 333 a.a) full-length human protein.

Sequence
MRIWWLLLAIEICTGNINSQDTCRQGHPGIPGNPGHNGLPGRDGRDGAKGDKGDAGEPGRPGSPGKDGTSGEKGERGADGKVEAKGIKGDQGSRGSPGKHGPKGLAGPMGEKGLRGETGPQGQKGNKGDVGPTGPEGPRGNIGPLGPTGLPGPMGPIGKPGPKGEAGPTGPQGEPGVRGIRGWKGDRGEKGKIGETLVLPKSAFTVGLTVLSKFPSSDVPIKFDKILYNEFNHYDTAAGKFTCHIAGVYYFTYHITVFSRNVQVSLVKNGVKILHTKDAYMSSEDQASGGIVLQLKLGDEVWLQVTGGERFNGLFADEDDDTTFTGFLLFSSP

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service