Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB1400745

Sigma-Aldrich

Anti-ACSS1 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ACAS2L, Anti-AceCS2L, Anti-MGC33843

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACSS1(84532)

General description

ACSS1 (acyl-CoA synthetase short chain family member 1) gene is mapped to human chromosome 20p11.21. The protein is highly expressed in heart and skeletal muscles.

Immunogen

ACSS1 (AAH39261.1, 1 a.a. ~ 689 a.a) full-length human protein.

Sequence
MAARTLGRGVGRLLGSLRGLSGQPARPPCGVSAPRRAASGPSGSAPAVAAAAAQPGSYPALSAQAAREPAAFWGPLARDTLVWDTPYHTVWDCDFSTGKIGWFLGGQLNVSVNCLDQHVRKSPESVALIWERDEPGTEVRITYRELLETTCRLANTLKRHGVHRGDRVAIYMPVSPLAVAAMLACARIGAVHTVIFAGFSAESLAGRINDAKCKVVITFNQGLRGGRVVELKKIVDEAVKHCPTVQHVLVAHRTDNKVHMGDLDVPLEQEMAKEDPVCAPESMGSEDMLFMLYTSGSTGMPKGIVHTQAGYLLYAALTHKLVFDHQPGDIFGCVADIGWITGHSYVVYGPLCNGATSVLFESTPVYPNAGRYWETVERLKINQFYGAPTAVRLLLKYGDAWVKKYDRSSLRTLGSVGEPINCEAWEWLHRVVGDSRCTLVDTWWQTETGGICIAPRPSEEGAEILPAMAMRPFFGIVPVLMDEKGSVMEGSNVSGALCISQAWPGMARTIYGDHQRFVDAYFKAYPGYYFTGDGAYRTEGGYYQITGRMDDVINISGHRLGTAEIEDAIADHPAVPESAVIGYPHDIKGEAAFAFIVVKDSAGDSDVVVQELKSMVATKIAKYAVPDEILVVKRLPKTRSGKVMRRLLRKIITSEAQELGDTTTLEDPSIIAEILSVYQKCKDKQAAAK

Biochem/physiol Actions

The enzyme coded by ACSS1 gene is responsible for the conversion for acetate to acetyl CoA in the mitochondria. It thereby contributes to energy production during ketosis, especially in the heart and skeletal muscle.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Fujino et al.
The Journal of biological chemistry, 276(14), 11420-11426 (2001-01-21)
Using peptide sequences derived from bovine cardiac acetyl-CoA synthetase (AceCS), we isolated and characterized cDNAs for a bovine and murine cardiac enzyme designated AceCS2. We also isolated a murine cDNA encoding a hepatic type enzyme, designated AceCS1, identical to one
Lei Jin et al.
The Journal of biological chemistry, 284(36), 24394-24405 (2009-06-19)
SIRT3 is a major mitochondrial NAD(+)-dependent protein deacetylase playing important roles in regulating mitochondrial metabolism and energy production and has been linked to the beneficial effects of exercise and caloric restriction. SIRT3 is emerging as a potential therapeutic target to
Mijin Yun et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(8), 1222-1228 (2009-07-21)
We analyzed the pattern of (11)C-acetate and (18)F-FDG uptake on PET/CT in patients with hepatocellular carcinoma (HCC). We also assessed the expression of important regulatory enzymes related to glycolysis and lipid synthesis in relation to (18)F-FDG and (11)C-acetate uptake in
Kazunori Kawaguchi et al.
The American journal of pathology, 186(8), 2055-2067 (2016-06-19)
Notch signaling abnormalities are reported to be involved in the acceleration of malignancy in solid tumors and stem cell formation or regeneration in various organs. We analyzed specific genes for DNA copy number variations in liver cancer cells and investigated
Bjoern Schwer et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(27), 10224-10229 (2006-06-22)
We report that human acetyl-CoA synthetase 2 (AceCS2) is a mitochondrial matrix protein. AceCS2 is reversibly acetylated at Lys-642 in the active site of the enzyme. The mitochondrial sirtuin SIRT3 interacts with AceCS2 and deacetylates Lys-642 both in vitro and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service