Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB1400618

Sigma-Aldrich

Monoclonal Anti-DDX43 antibody produced in mouse

clone 3G12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZp434H2114, Anti-HAGE

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$673.00

$673.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$673.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$673.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDX43(55510)

General description

The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression. (provided by RefSeq)

Immunogen

DDX43 (NP_061135.1, 2 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SHHGGAPKASTWVVASRRSSTVSRAPERRPAEELNRTGPEGYSVGRGGRWRGTSRPPEAVAAGHEELPLCFALKSHFVGAVIGRGGSKI

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam J Linley et al.
The Journal of biological chemistry, 287(17), 13633-13643 (2012-03-07)
Malignant melanoma-initiating cells (MMIC) are a subpopulation of cells responsible for melanoma tumor growth and progression. They are defined by the expression of the ATP-binding cassette (ABC) subfamily B member 5 (ABCB5). Here, we identified a critical role for the
Tanu Talwar et al.
The Journal of biological chemistry, 292(25), 10429-10443 (2017-05-05)
The K-homology (KH) domain is a nucleic acid-binding domain present in many proteins but has not been reported in helicases. DDX43, also known as HAGE (helicase antigen gene), is a member of the DEAD-box protein family. It contains a helicase
M G Mathieu et al.
Cell death & disease, 5, e1061-e1061 (2014-02-15)
The tumour suppressor PML (promyelocytic leukaemia protein) regulates several cellular pathways involving cell growth, apoptosis, differentiation and senescence. PML also has an important role in the regulation of stem cell proliferation and differentiation. Here, we show the involvement of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service