Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1400266

Sigma-Aldrich

Anti-ST13 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-AAG2, Anti-FAM10A1, Anti-HIP, Anti-HOP, Anti-HSPABP, Anti-HSPABP1, Anti-MGC129952, Anti-P48, Anti-PRO0786, Anti-SNC6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ST13(6767)

General description

The protein encoded by this gene is an adaptor protein that mediates the association of the heat shock proteins HSP70 and HSP90. This protein has been shown to be involved in the assembly process of glucocorticoid receptor, which requires the assistance of multiple molecular chaperones. The expression of this gene is reported to be downregulated in colorectal carcinoma tissue suggesting that is a candidate tumor suppressor gene. (provided by RefSeq)

Immunogen

ST13 (NP_003923.2, 1 a.a. ~ 369 a.a) full-length human protein.

Sequence
MDPRKVNELRAFVKMCKQDPSVLHTEEMRFLREWVESMGGKVPPATQKAKSEENTKEEKPDSKKVEEDLKADEPSSEESDLEIDKEGVIEPDTDAPQEMGDENAEITEEMMDQANDKKVAAIEALNDGELQKAIDLFTDAIKLNPRLAILYAKRASVFVKLQKPNAAIRDCDRAIEINPDSAQPYKWRGKAHRLLGHWEEAAHDLALACKLDYDEDASAMLKEVQPRAQKIAEHRRKYERKREEREIKERIERVKKAREEHERAQREEEARRQSGAQYGSFPGGFPGGMPGNFPGGMPGMGGGMPGMAGMPGLNEILSDPEVLAAMQDPEVMVAFQDVAQNPANMSKYQSNPKVMNLISKLSAKFGGQA

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service