Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1400213

Sigma-Aldrich

Anti-PLOD2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-LH2, Anti-TLH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLOD2(5352)

General description

The protein encoded by this gene is a membrane-bound homodimeric enzyme that is localized to the cisternae of the rough endoplasmic reticulum. The enzyme (cofactors iron and ascorbate) catalyzes the hydroxylation of lysyl residues in collagen-like peptides. The resultant hydroxylysyl groups are attachment sites for carbohydrates in collagen and thus are critical for the stability of intermolecular crosslinks. Some patients with Ehlers-Danlos syndrome type VIB have deficiencies in lysyl hydroxylase activity. Mutations in the coding region of this gene are associated with Bruck syndrome. Alternative splicing results in multiple transcript variants encoding different isoforms. (provided by RefSeq)

Immunogen

PLOD2 (NP_891988.1, 1 a.a. ~ 758 a.a) full-length human protein.

Sequence
MGGCTVKPQLLLLALVLHPWNPCLGADSEKPSSIPTDKLLVITVATKESDGFHRFMQSAKYFNYTVKVLGQGEEWRGGDGINSIGGGQKVRLMKEVMEHYADQDDLVVMFTECFDVIFAGGPEEVLKKFQKANHKVVFAADGILWPDKRLADKYPVVHIGKRYLNSGGFIGYAPYVNRIVQQWNLQDNDDDQLFYTKVYIDPLKREAINITLDHKCKIFQTLNGAVDEVVLKFENGKARAKNTFYETLPVAINGNGPTKILLNYFGNYVPNSWTQDNGCTLCEFDTVDLSAVDVHPNVSIGVFIEQPTPFLPRFLDILLTLDYPKEALKLFIHNKEVYHEKDIKVFFDKAKHEIKTIKIVGPEENLSQAEARNMGMDFCRQDEKCDYYFSVDADVVLTNPRTLKILIEQNRKIIAPLVTRHGKLWSNFWGALSPDGYYARSEDYVDIVQGNRVGVWNVPYMANVYLIKGKTLRSEMNERNYFVRDKLDPDMALCRNAREMTLQREKDSPTPETFQMLSPPKGVFMYISNRHEFGRLLSTANYNTSHYNNDLWQIFENPVDWKEKYINRDYSKIFTENIVEQPCPDVFWFPIFSEKACDELVEEMEHYGKWSGGKHHDSRISGGYENVPTDDIHMKQVDLENVWLHFIREFIAPVTLKVFAGYYTKGFALLNFVVKYSPERQRSLRPHHDASTFTINIALNNVGEDFQGGGCKFLRYNCSIESPRKGWSFMHPGRLTHLHEGLPVKNGTRYIAVSFIDP

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masum M Mia et al.
Cell and tissue research, 363(3), 775-789 (2015-10-11)
Fibrosis is a chronic disorder affecting many organs. A universal process in fibrosis is the formation of myofibroblasts and the subsequent collagen deposition by these cells. Transforming growth factor beta1 (TGFβ1) plays a major role in the formation of myofibroblasts
Rutger A F Gjaltema et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(26), 7142-7147 (2016-06-15)
Collagens are subjected to extensive posttranslational modifications, such as lysine hydroxylation. Bruck syndrome (BS) is a connective tissue disorder characterized at the molecular level by a loss of telopeptide lysine hydroxylation, resulting in reduced collagen pyridinoline cross-linking. BS results from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service