Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1400195

Sigma-Aldrich

Anti-PCSK2 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-NEC2, Anti-PC2, Anti-SPC2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCSK2(5126)

General description

The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a proinsulin-processing enzyme that plays a key role in regulating insulin biosynthesis. It is also known to cleave proopiomelanocortin, proenkephalin, prodynorphin and proluteinizing-hormone-releasing hormone. The use of alternate polyadenylation sites has been found for this gene. (provided by RefSeq)

Immunogen

PCSK2 (NP_002585.2, 1 a.a. ~ 638 a.a) full-length human protein.

Sequence
MKGGCVSQWKAAAGFLFCVMVFASAERPVFTNHFLVELHKGGEDKARQVAAEHGFGVRKLPFAEGLYHFYHNGLAKAKRRRSLHHKQQLERDPRVKMALQQEGFDRKKRGYRDINEIDINMNDPLFTKQWYLINTGQADGTPGLDLNVAEAWELGYTGKGVTIGIMDDGIDYLHPDLASNYNAEASYDFSSNDPYPYPRYTDDWFNSHGTRCAGEVSAAANNNICGVGVAYNSKVAGIRMLDQPFMTDIIEASSISHMPQLIDIYSASWGPTDNGKTVDGPRELTLQAMADGVNKGRGGKGSIYVWASGDGGSYDDCNCDGYASSMWTISINSAINDGRTALYDESCSSTLASTFSNGRKRNPEAGVATTDLYGNCTLRHSGTSAAAPEAAGVFALALEANLGLTWRDMQHLTVLTSKRNQLHDEVHQWRRNGVGLEFNHLFGYGVLDAGAMVKMAKDWKTVPERFHCVGGSVQDPEKIPSTGKLVLTLTTDACEGKENFVRYLEHVQAVITVNATRRGDLNINMTSPMGTKSILLSRRPRDDDSKVGFDKWPFMTTHTWGEDARGTWTLELGFVGSAPQKGVLKEWTLMLHGTQSAPYIDQVVRDYQSKLAMSKKEELEEELDEAVERSLKSILNKN

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service