Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB1306673

Sigma-Aldrich

ANTI-LC3 (APG8A) (N-TERM) antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, MAP1 light chain 3-like protein 1, MAP1LC3A, Microtubule-associated proteins 1A/1B light chain 3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

14272 Da

species reactivity

human, mouse, rat

technique(s)

immunofluorescence: 1:200
immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Zhang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 37(4), 1442-1453 (2015-10-23)
There have been many studies on the etiology of osteoarthritis (OA) with regard to the function of inflammatory cytokines, the process of cartilage degradation, the function of miR-146a, hypoxia stimulation and autophagy in OA chondrocytes, but there have been no
Xiaowen Yu et al.
Experimental and therapeutic medicine, 16(3), 2219-2228 (2018-09-07)
Strokes are a major cause of neurological disability. Stem cell replacement therapy is a potential novel strategy of treating patients that have experienced strokes. The present study examined the protective role of neural stem cell (NSC) administration in oxygen-glucose deprivation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service