Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1305952

Sigma-Aldrich

ANTI-TIAM1(C-TERMINAL) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

T-lymphoma invasion and metastasis-inducing protein 1, TIAM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

177508 Da

species reactivity

human

technique(s)

western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIAM1(7074)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Izumi et al.
Cell death & disease, 10(4), 267-267 (2019-03-21)
Accumulating evidence suggests that cancer cells with stem cell-like features have higher resistance to chemotherapeutic agents. Herein, we identified T-lymphoma invasion and metastasis-inducing protein-1 (TIAM1) as one of the Wnt-signaling associated genes which drives self-renewal and its expression is upregulated
Xiaoping Zhang et al.
Oncogene, 38(5), 699-715 (2018-09-02)
Anaplastic thyroid cancer (ATC) is associated with poor prognosis and is often untreatable. MicroRNA 483-3p (miR-483) and partitioning-defective 3 (Pard3), a member of the Pard family, have functions and regulatory mechanisms in ATC. The abnormal regulation of miR-483 may play
Hidetsugu Kawai et al.
Neoplasia (New York, N.Y.), 21(8), 788-801 (2019-07-06)
Hematological malignancies harboring various ABL1 fusions are expected to be sensitive to tyrosine kinase inhibitors (TKIs), similar to those with BCR-ABL1. However, SEPT9-ABL1 exhibits TKI resistance both in vitro and in vivo. SEPT9-ABL1 has the same ABL1 region as seen
Takeo Saneyoshi et al.
Neuron, 102(6), 1199-1210 (2019-05-13)
Long-term synaptic plasticity requires a mechanism that converts short Ca2+ pulses into persistent biochemical signaling to maintain changes in the synaptic structure and function. Here, we present a novel mechanism of a positive feedback loop, formed by a reciprocally activating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service