Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1305776

Sigma-Aldrich

ANTI-LC3 (APG8B) (T6) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, MAP1ALC3, MAP1LC3B, Microtubule-associated proteins 1A/1B light chain 3B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

14688 Da

species reactivity

human, mouse

technique(s)

western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

LC3B or MAP1LC3B (microtubule associated protein 1 light chain 3 β) is a homolog of the ATG8 gene, which is an autophagy-related gene in yeast. The encoded protein is called pro-MAP1LC3B, a pro-form that is cleaved by ATG4 to produce a cytosolic form called MAP1LC3B-I.

Biochem/physiol Actions

MAP1LC3B-I (microtubule associated protein 1 light chain 3 β-I) associates with MAP1LC3B-II before being inserted into the inner and outer membranes of the growing autophagic vesicle during autophagy. The cellular distribution and processing of MAP1LC3B facilitates monitoring of autophagy. Cells that lack MAP1LC3B are found to be defective in the formation of autophagic vesicles, with altered phenotypes associated with the role of autophagy in energy homeostasis and removal of protein aggregates. MAP1LC3B can serve as an autophagic marker.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Autophagic marker MAP1LC3B expression levels are associated with carotid atherosclerosis symptomatology.
Swaminathan B, et al.
PLoS ONE (2014)
The unfolded protein response protects human tumor cells during hypoxia through regulation of the autophagy genes MAP1LC3B and ATG5.
Rouschop KM, et al.
The Journal of Clinical Investigation (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service