Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1305567

Sigma-Aldrich

MONOCLONAL ANTI-BETA-ACTIN antibody produced in mouse

clone 8H10D10, crude ascites, buffered aqueous solution

Synonym(s):

ACTB, Actin, cytoplasmic 1, Actin, cytoplasmic 1, N-terminally processed, Beta-actin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

crude ascites

antibody product type

primary antibodies

clone

8H10D10, monoclonal

form

buffered aqueous solution

mol wt

41737 Da

species reactivity

human

technique(s)

immunohistochemistry: 1:10-1:50
western blot: 1:5000-1:20000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTB(60)

General description

ACTB (actin β) gene codes for β-actin. It is a cytoskeletal housekeeping protein. It is present in the cytoplasm. This gene is located on human chromosome 7p22.1.

Application

MONOCLONAL ANTI-BETA-ACTIN antibody has been used in western blotting.

Biochem/physiol Actions

β-Actin modulates cell growth, migration and the G-actin pool. Mutations in ACTB leads to pleiotropic developmental disorder.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radiation alters the cargo of exosomes released from squamous head and neck cancer cells to promote migration of recipient cells.
Mutschelknaus L, et al.
Scientific Reports, 7(1), 12423-12423 (2017)
Lisa Mutschelknaus et al.
Scientific reports, 7(1), 12423-12423 (2017-10-01)
Radiation is a highly efficient therapy in squamous head and neck carcinoma (HNSCC) treatment. However, local recurrence and metastasis are common complications. Recent evidence shows that cancer-cell-derived exosomes modify tumour cell movement and metastasis. In this study, we link radiation-induced
Gabriela M Wright et al.
PloS one, 16(6), e0252927-e0252927 (2021-06-18)
Theoretically, small molecule CDK4/6 inhibitors (CDK4/6is) represent a logical therapeutic option in non-small cell lung cancers since most of these malignancies have wildtype RB, the key target of CDKs and master regulator of the cell cycle. Unfortunately, CDK4/6is are found
Lisa Mutschelknaus et al.
PloS one, 11(3), e0152213-e0152213 (2016-03-24)
Exosomes are nanometer-sized extracellular vesicles that are believed to function as intercellular communicators. Here, we report that exosomes are able to modify the radiation response of the head and neck cancer cell lines BHY and FaDu. Exosomes were isolated from
Mengmeng Fan et al.
Oncology reports, 47(1) (2021-11-06)
The epithelial‑stromal interaction 1 gene (EPSTI1) is known to play multiple roles in the malignant progression of breast cancer and also in some aspects of the immune responses to the tumor. However, the relevance of the gene in the onset/progression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service