Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300975

Sigma-Aldrich

Anti-ULK3 (center) antibody produced in rabbit

Ig fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Serine/threonine-protein kinase ULK3, Anti-Unc-51-like kinase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ULK3(25989)

General description

ULK3 is a member of the ULK kinase family, one of three human homologs of the yeast autophagy-specific kinase.

Immunogen

ULK3 (NP_001092906, 92-126)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human ULK3.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service