Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300869

Sigma-Aldrich

Anti-THPO (C-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-C-mpl ligand, Anti-ML, Anti-Megakaryocyte colony-stimulating factor, Anti-Thrombopoietin, Myeloproliferative leukemia virus oncogene ligand

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... THPO(7066)

General description

Megakaryocytopoiesis is the cellular development process that leads to platelet production. THPO is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. It is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene. It is a lineage specific growth factor, produced in the liver, kidney and skeletal muscle.[1] It is an acidic glycoprotein. The gene encoding it is localized on human chromosome 3q27.1.

Immunogen

THPO (NP_000451, 305-339)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide at the C-term of human THPO.

Biochem/physiol Actions

Thrombopoietin (TPO) stimulates the proliferation and maturation of megakaryocytes, and promotes increased circulating levels of platelets in vivo. TPO signals through the cellular proto-oncogene (c-mpl) receptor and acts as an important regulator of circulating platelets. High levels of TPO have been observed in patients with aplastic anemia. Human and murine TPO exhibits cross-species reactivity.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service