Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300845

Sigma-Aldrich

Anti-PARG (N-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-Poly(ADP-ribose) glycohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARG(8505)

General description

PARG (Poly ADP-ribose glycohydrolase) is a catabolic enzyme with molecular mass ranging from 60-74kDa. It has four isoforms namely G1, G2, G3 and G4 with high protease sensitivity.

Immunogen

PARG (NP_003622, 329-364)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human Parg.

Application

Anti-PARG (N-term) antibody produced in rabbit is suitable for indirect ELISA (1:1000) and immunohistochemistry (1:50-1:100) applications.

Biochem/physiol Actions

PARG (Poly ADP-ribose glycohydrolase) is involved in several gene regulation processes such as maintenance of chromatin structure, transcription, DNA repair and apoptosis. It regulates retinoic acid receptor (RAR)-mediated transcription during chromatin remodeling. It catabolically separate ADP-ribose polymers synthesized by poly-(ADP-ribose) polymerases. A study shows that PARG plays a therapeutic role in various physiological conditions such as inflammation, ischemia, stroke, and cancer chemotherapy.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wookee Min et al.
Frontiers in bioscience (Landmark edition), 14, 1619-1626 (2009-03-11)
Poly (ADP-robose) glycohydrolase (PARG) is a catabolic enzyme that cleaves ADP-ribose polymers synthesized by members of the poly (ADP-ribose) polymerase (PARP) family of enzymes. The growing evidence supports the importance of a tight control of poly (ADP-ribose) metabolism by the
M-E Bonicalzi et al.
Cellular and molecular life sciences : CMLS, 62(7-8), 739-750 (2005-05-04)
Poly(ADP-ribose) glycohydrolase (PARG) is a catabolic enzyme that cleaves ADP-ribose polymers formed by members of the PARP family of enzymes. Despite its discovery and subsequent partial purification in the 1970s and the cloning of its single gene in the late
Nicolas Le May et al.
Molecular cell, 48(5), 785-798 (2012-10-30)
Poly-(ADP-ribose) glycohydrolase (PARG) is a catabolic enzyme that cleaves ADP-ribose polymers synthesized by poly-(ADP-ribose) polymerases. Here, transcriptome profiling and differentiation assay revealed a requirement of PARG for retinoic acid receptor (RAR)-mediated transcription. Mechanistically, PARG accumulates early at promoters of RAR-responsive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service