Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300716

Sigma-Aldrich

Anti-CDA (C-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-CDD, Anti-Cytidine aminohydrolase, Anti-Cytidine deaminase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$603.00

$603.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$603.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$603.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDA(978)

General description

CDA forms a homotetramer that catalyzes the irreversible hydrolytic deamination of cytidine and deoxycytidine to uridine and deoxyuridine, respectively. This protein is one of several deaminases responsible for maintaining the cellular pyrimidine pool. Mutations in the CDA gene are associated with decreased sensitivity to the cytosine nucleoside analogue cytosine arabinoside used in the treatment of certain childhood leukemias.

Immunogen

CDA (NP_001776, 123-156)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-term region of human CDA.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woosuk Kim et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(15), 4027-4032 (2016-04-02)
Deoxycytidine kinase (dCK), a rate-limiting enzyme in the cytosolic deoxyribonucleoside (dN) salvage pathway, is an important therapeutic and positron emission tomography (PET) imaging target in cancer. PET probes for dCK have been developed and are effective in mice but have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service