Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300641

Sigma-Aldrich

Anti-UBE2V1 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DJ1185N5.1.3, Anti-TRAF6-regulated IKK activator 1 β Uev1A, Anti-Ubiquitin-conjugating enzyme E2 variant 1 (Isoform 3, CROC-1A), Ubiquitin carrier protein, Ubiquitin-protein ligase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$603.00

$603.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$603.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$603.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE2V1(7335)

General description

The CROC1 isoforms, also known as UBE2V1, show sequence similarity to ubiquitin-conjugating enzymes (UBCs, or E2s) but lack the conserved cysteine residue critical to catalytic activity of E2s.1 Northern blot analysis detected approximately 2.1- and 2.5-kb CROC1 transcripts in all human tissues examined, with the highest levels in brain, skeletal muscle, and kidney. Partial human intestinal epithelial cell cDNAs have been isolated containing the 3-primecoding sequence and 3-prime untranslated region of UBE2V1, also called UEV1.2 UEV1 does not have ubiquitin-conjugating activity in vitro. UEV1 transcripts are downregulated upon differentiation of a colon carcinoma cell line.1 Constitutive expression of exogenous UEV1 protein in these colon carcinoma cells inhibits their capacity to differentiate upon confluence and induces changes in cell cycle behavior associated with inhibition of CDK1. A heterodimeric protein complex has been identified that links TRAF6 to IKK activation.3 Peptide mass fingerprinting analysis revealed that this complex is composed of the ubiquitin conjugating enzyme UBC13 and the UBC-like protein UBE2V1, also called UEV1A. TRAF6, a RING domain protein, functions together with UBC13/UEV1A to catalyze the synthesis of unique polyubiquitin chains linked through lysine-63 (K63) of ubiquitin. Blockade of this polyubiquitin chain synthesis, but not inhibition of the proteasome, prevents the activation of IKK by TRAF6. These results unveil a new regulatory function for ubiquitin, in which IKK is activated through the assembly of K63-linked polyubiquitin chains.

The previously assigned protein identifier Q9GZW1 has beenmerged into Q13404. Full details can be found on the UniProt database.

Immunogen

UBE2V1 (NP_068823, 141-187)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human UBE2V1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service