Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300556

Sigma-Aldrich

Anti-NR6A1 (C-term) antibody produced in rabbit

Ig fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-GCNF, Anti-Germ cell nuclear factor, Anti-HGCNF, Anti-RTR, Retinoid receptor-related testis-specific receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR6A1(2649)

Related Categories

General description

NR6A1 has been shown to affect gene expression in embryogenesis and spermatogenesis and is essential for embryonic survival and normal development. Loss of NR6A1 causes embryonic lethality and disrupts normal somitogenesis as well as neural-tube and axis formation. NR6A1 binds as a homodimer to the DR0 DNA response element and has been shown to regulate transcription of genes including protamines 1 and 2 and Oct4. NR6A1 repression function has been shown to be mediated by interaction with the co-repressors N-COR and SMRT in the absence of ligand.

Immunogen

NR6A1 (NP_201591, 340-375)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human NR6A1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service