Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300551

Sigma-Aldrich

Anti-ERAS (N-term) antibody produced in rabbit

Ig fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-E-Ras, Anti-Embryonic stem cell-expressed Ras

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
indirect immunofluorescence: 1:10-1:50
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERAS(3266)

General description

Ras proteins bind GDP/GTP and possess intrinsic GTPase activity. Point mutations of several amino acids of human RAS, including gly12, ala59, or glu63, render the protein constitutively active. Embryonic stem cell-expressed Ras (ERAS) has serine, alanine, and asparagine at the positions corresponding to gly12, ala59, and glu63 of human RAS, suggesting that it is constitutively active.The PI3K (phosphoinositide 3-kinase) pathway is important for proliferation, survival and maintenance of pluripotency in ES cells. The PI3K pathway is activated by growth factors and cytokines including insulin and leukaemia inhibitory factor. In addition to these exogenous factors, the PI3K pathway is endogenously activated by the constitutively active Ras family protein ERas (ES cell-expressed Ras). ERas null ES cells maintained pluripotency but show significantly reduced growth and tumorigenicity, which can be rescued by expression of ERas cDNA or by activated phosphatidylinositol 3-hydroxykinase. The transforming oncogene ERAS appears to be important in the tumor-like growth properties of ES cells. This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human ERAS.

Immunogen

ERAS (NP_853510, 18-52)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human ERAS.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service